missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl chloroformate, 97%

Product Code. 10514284 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 10514284

Brand: Thermo Scientific Acros 170805000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7693-46-1 | C7H4ClNO4 | 201.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7693-46-1
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.57
MDL Number MFCD00007321
InChI Key NXLNNXIXOYSCMB-UHFFFAOYSA-N
Synonym 4-nitrophenyl chloroformate, carbonochloridic acid, 4-nitrophenyl ester, chloroformic acid 4-nitrophenyl ester, p-nitrophenyl chloroformate, chloroformic acid p-nitrophenyl ester, 4-nitrophenyl carbonochloridate, formic acid, chloro-, p-nitrophenyl ester, 4-nitrophenylchlorocarbonat, 4-nitrophenyl choroformate
PubChem CID 82129
IUPAC Name (4-nitrophenyl) carbonochloridate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 73.0°C to 81.0°C
Color Beige-Yellow to White
Boiling Point 159.0°C to 162.0°C (19.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula ClCO2C6H4NO2
Quantity 500 g
Beilstein 06, I, 120
Fieser 02,297
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in toluene and benzene
Formula Weight 201.57
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 4-Nitrophenyl chloroformate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.