Learn More
4-Vinylbenzeneboronic acid, 97%
Description
4-Vinylbenzeneboronic acid is widely used in Suzuki reaction. It is also used in the preparation of boronic acid containing polymers, which is used in potential biomedical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Vinylbenzeneboronic acid is widely used in Suzuki reaction. It is also used in the preparation of boronic acid containing polymers, which is used in potential biomedical applications.
Solubility
Soluble in ethanol, dimethylsulfoxide, tetrahydrofuran and diethyl ether. Slightly soluble in water.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∼190°C |
| Quantity | 5 g |
| Beilstein | 2829387 |
| Solubility Information | Soluble in ethanol,dimethylsulfoxide,tetrahydrofuran and diethyl ether. Slightly soluble in water. |
| Formula Weight | 147.97 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Vinylbenzeneboronic acid |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.