Learn More
5-(2-Phenyleth-1-ynyl)-2-furoic acid, Tech., Thermo Scientific™ Product Code.: 10652895
5-(2-Phenyleth-1-ynyl)-2-furoic acid, Tech., Thermo Scientific™
Product Code. 10652895
10 g
Click to view available options
Quantity:
250 mg
1 g
10 g
Product Code. 10652895
Brand: Thermo Scientific Maybridge KM08625EA
Request Bulk or Custom Format
Chemical Identifiers
CAS 130423-83-5
Molecular Formula C13H8O3
Molecular Weight (g/mol) 212.20
MDL Number MFCD00541949
InChI Key HFYACYZCOUYIFR-UHFFFAOYSA-N
Synonym 5-phenylethynyl furan-2-carboxylic acid, 5-2-phenyleth-1-ynyl-2-furoic acid, 5-2-phenylethynyl furan-2-carboxylic acid, 5-phenylethynyl-2-furoic acid, 2-furancarboxylic acid,5-2-phenylethynyl, maybridge1_008982, acmc-20a65w, 5-phenylethynyl-2-furoic acid, tech, 5-phenylethynyl-furan 2 carboxylic acid
PubChem CID 2737138
IUPAC Name 5-(2-phenylethynyl)furan-2-carboxylic acid
SMILES OC(=O)C1=CC=C(O1)C#CC1=CC=CC=C1
Specifications
Assay Percent Range 95%
Quantity 10 g
Formula Weight 212.2
Percent Purity Tech.
Grade Technical
Chemical Name or Material 5-(2-Phenyleth-1-ynyl)-2-furoic acid
Product Content Correction
Product Title
5-(2-Phenyleth-1-ynyl)-2-furoic acid, Tech., Thermo Scientific™ >