missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Hydroxyphenyl)pentanoic acid, 98%, Thermo Scientific™

Product Code. 11466137
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11466137

Brand: Thermo Scientific Alfa Aesar B25107.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-(4-Hydroxyphenyl)pentanoic acid is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4654-08-4

Specifications

Melting Point 116°C to 120°C
Molecular Formula C11H14O3
MDL Number MFCD04039777
Quantity 1 g
Synonym 5-4-hydroxyphenyl pentanoic acid, 5-4-hydroxyphenyl valeric acid, pubchem16852, acmc-20am8d, 5-4-hydroxyphenyl pentanoicacid, benzenepentanoic acid, 4-hydroxy
Solubility Information Soluble in water.
InChI Key YSSJQFONKASLKM-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CCCCC(=O)O)O
IUPAC Name 5-(4-hydroxyphenyl)pentanoic acid
Molecular Weight (g/mol) 194.23
PubChem CID 7016423
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material 5-(4-Hydroxyphenyl)pentanoic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.