missing translation for 'onlineSavingsMsg'
Learn More

5-[5-(2-Thienyl)-2-thienyl]thiophene-2-carbaldehyde, Thermo Scientific™

Product Code. p-4813172
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10577363

Brand: Thermo Scientific Maybridge SEW04053CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7342-41-8
Molecular Formula C13H8OS3
Molecular Weight (g/mol) 276.39
MDL Number MFCD00115181
InChI Key PMPDDPJYARBNGV-UHFFFAOYSA-N
Synonym 2,2':5',2-terthiophene-5-carboxaldehyde, 5-5-2-thienyl-2-thienyl thiophene-2-carbaldehyde, 2,2':5',2-terthiophene-5-carbaldehyde, 5-formyl-2,2':5',2-terthiophene, chembl91933, 5-5-thiophen-2-ylthiophen-2-yl thiophene-2-carbaldehyde, 5-5-thiophen-2-yl thiophen-2-yl thiophene-2-carbaldehyde, 2-formyl-alpha-terthienyl
PubChem CID 454742
SMILES O=CC1=CC=C(S1)C1=CC=C(S1)C1=CC=CS1

Specifications

Quantity 250 mg
IUPAC Name 5-(5-thiophen-2-ylthiophen-2-yl)thiophene-2-carbaldehyde
Formula Weight 276.4
Chemical Name or Material 5-[5-(2-Thienyl)-2-thienyl]thiophene-2-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.