missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-methylbenzonitrile, 97%, Thermo Scientific Chemicals

Product Code. p-7053762
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15477958

Brand: Thermo Scientific Alfa Aesar H61422.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50670-64-9
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.166
MDL Number MFCD00017608
InChI Key YDZVQWCVKXYGIU-UHFFFAOYSA-N
Synonym 5-amino-2-methylbenzenecarbonitrile, 2-methyl-5-aminobenzonitrile, 4-amino-2-cyanotoluene, 3-cyano-4-methylaniline, 5-amino-2-methyl-benzonitrile, benzonitrile, 5-amino-2-methyl, 5-amino-o-tolunitrile, acmc-20a01k, aminomethylbenzenecarbonitrile, 3-amino-6-methylbenzonitrile
PubChem CID 2735365
IUPAC Name 5-amino-2-methylbenzonitrile
SMILES CC1=C(C=C(C=C1)N)C#N

Specifications

Melting Point 88°C to 91°C
Quantity 5 g
UN Number UN3439
Formula Weight 132.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Amino-2-methylbenzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.