missing translation for 'onlineSavingsMsg'
Learn More

5-Bromovaleryl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11449186
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

5-Bromovaleryl chloride was used in the preparation of aromatic azides and organic gelators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromovaleryl chloride was used in the preparation of aromatic azides and organic gelators.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from water/ moisture, oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4509-90-4
Molecular Formula C14H11N3O2
Molecular Weight (g/mol) 253.26
MDL Number MFCD00013660
InChI Key QANQQXSQVQYQFI-UHFFFAOYSA-N
Synonym 5-bromovaleryl chloride, 5-bromovaleroyl chloride, 5-bromo-pentanoyl chloride, 5-bromovaleric acid chloride, 5-bromovalerylchloride, bromovalerylchloride, bromovaleryl chloride, 5-bromovaleric chloride, 5-bromopentanoylchloride, 5-bromo pentanoyl chloride
PubChem CID 521173
IUPAC Name 5-bromopentanoyl chloride
SMILES CC(=O)NC1=CC=C2OC(=NC2=C1)C1=CC=NC=C1

Specifications

Density 1.49
Boiling Point 43°C to 45°C (0.2 mmHg)
Flash Point 107°C (225°F)
Odor Irritating
Linear Formula Br(CH2)4COCl
Refractive Index 1.491
Quantity 5 g
UN Number UN3265
Beilstein 1745663
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 199.46
Percent Purity 98%
Chemical Name or Material 5-Bromovaleryl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'