missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methoxybenzylamine, JRD, 97%, Thermo Scientific™

Product Code. p-7031724
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15455667

Brand: Thermo Scientific Alfa Aesar H32897.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148870-38-6
Molecular Formula C8H10FNO
Molecular Weight (g/mol) 155.17
MDL Number MFCD06213625
InChI Key IYQIJUZZRLPXIL-UHFFFAOYSA-N
Synonym 5-fluoro-2-methoxybenzylamine, benzenemethanamine,5-fluoro-2-methoxy, 1-5-fluoro-2-methoxyphenyl methanamine, 5-fluoro-2-methoxyphenyl methanamine, acmc-20n5ju, 5-fluoro-2-methoxybenzyl amine, 5-fluoro-2-methoxybenzylamine, jrd, 5-fluoro-2-methoxy-benzenemethanamine, 5-fluoro-2-methoxyphenyl methylamine, 5-fluoro-2-methoxyphenyl-methanamine
PubChem CID 18348829
IUPAC Name (5-fluoro-2-methoxyphenyl)methanamine
SMILES COC1=C(CN)C=C(F)C=C1

Specifications

Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 155.17
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-methoxybenzylamine, JRD
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.