missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-7-methylindole, 97%

Product Code. p-7054726
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15449638

Brand: Thermo Scientific Alfa Aesar H63123.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 61019-05-4 | C10H11NO | 161.204 g/mol

5-Methoxy-7-methylindole is employed as a intermediate for chemical research and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methoxy-7-methylindole is employed as a intermediate for chemical research and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61019-05-4
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD09027022
InChI Key YGPVRHHGKCQSIL-UHFFFAOYSA-N
Synonym 5-methoxy-7-methylindole, 1h-indole, 5-methoxy-7-methyl, ksc612k7l, 5-methoxy7-methyl-1h-indole
PubChem CID 12338917
IUPAC Name 5-methoxy-7-methyl-1H-indole
SMILES CC1=C2C(=CC(=C1)OC)C=CN2

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 161.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Methoxy-7-methylindole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.