missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-(tri-n-butylstannyl)pyridine, 96%, Thermo Scientific™

Product Code. 15428087
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15428087

Brand: Thermo Scientific Alfa Aesar H51377.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Methyl-2-(tri-n-butylstannyl)pyridine is used in Stille cross coupling reactions.

Solubility
Not miscible or difficult to mix.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 189195-41-3
Molecular Formula C18H33NSn
Molecular Weight (g/mol) 382.179
MDL Number MFCD07787402
InChI Key MVOHAZAWWAXIDR-UHFFFAOYSA-N
Synonym 5-methyl-2-tributylstannyl pyridine, 2-tributylstannyl-5-methylpyridine, pyridine,5-methyl-2-tributylstannyl, 5-methyl-2-tri-n-butylstannyl pyridine, acmc-209esb, 5-methyl-2-tributylstannylpyridine, 5-methy-2-tributylstannyl-pyridine, 5-methyl-2-tri-n-butylstannylpyridine, 5-methylpyridin-2-yl tributylstannane, 5-methyl-2-tributylstannyl-pyridine
PubChem CID 11132711
IUPAC Name tributyl-(5-methylpyridin-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=NC=C(C=C1)C

Specifications

Quantity 250 mg
UN Number UN2788
Solubility Information Not miscible or difficult to mix.
Formula Weight 382.16
Percent Purity 96%
Chemical Name or Material 5-Methyl-2-(tri-n-butylstannyl)pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.