missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-nitrohexahydropyrimidine-2,4,6-trione hydrate, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
50 g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 50 g |
IUPAC Name | 5-nitro-1,3-diazinane-2,4,6-trione;hydrate |
Formula Weight | 191.1 |
Percent Purity | 97% |
Chemical Name or Material | 5-Nitrohexahydropyrimidine-2, 4, 6-trione hydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction