missing translation for 'onlineSavingsMsg'
Learn More

5-O-tert-Butyldimethylsilyl-2,3-O-isopropylidene-alpha,beta-D-ribofuranose, Thermo Scientific™

34.26 EUR - 93.23 EUR

Chemical Identifiers

CAS 68703-51-5
Molecular Formula C14H28O5Si
Molecular Weight (g/mol) 304.458
MDL Number MFCD01075716
InChI Key JJLRGSYXWAKGJM-SGUBAKSOSA-N
Synonym 5-o-tert-butyldimethylsilyl-2,3-o-isopropylidene-d-ribose, 3as,4r,6r-6-tert-butyldimethylsilyl oxy methyl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-olShow MoreShow Less
PubChem CID 71314364
IUPAC Name (3aS,4R,6R)-6-[[tert-butyl(dimethyl)silyl]oxymethyl]-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-ol
SMILES CC1(OC2C(OC(C2O1)O)CO[Si](C)(C)C(C)(C)C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11342827
SDS View Documents
Thermo Scientific Alfa Aesar
L15986.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11332827
SDS View Documents
Thermo Scientific Alfa Aesar
L15986.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-O-tert-Butyldimethylsilyl-2,3-O-isopropylidene-α,β-D-ribofuranose is used as a starting material for the preparation of N-benzyl alfa-L-manno aminocyclopentitol , 4-deoxy-N-benzyl-alfa-L-manno aminocyclopentitol and N-benzyl 6,7-di-epi-trehalamine using stereoselective allylation. It is also used in oligosaccharides.

Solubility
Soluble in dichloromethane and ethyl acetate.

Notes
Incompatible with oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-O-tert-Butyldimethylsilyl-2,3-O-isopropylidene-alpha,beta-D-ribofuranose, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.