missing translation for 'onlineSavingsMsg'
Learn More

5-(Tri-n-butylstannyl)pyrimidine, 96%, Thermo Scientific Chemicals

79.60 EUR - 246.00 EUR

Chemical Identifiers

CAS 144173-85-3
Molecular Formula C16H30N2Sn
Molecular Weight (g/mol) 369.14
MDL Number MFCD01319056
InChI Key QRDQHTJNKPXXRQ-UHFFFAOYSA-N
Synonym 5-tributylstannyl pyrimidine, pyrimidine, 5-tributylstannyl, tributyl pyrimidin-5-yl stannane, 5-tri-n-butylstannyl pyrimidine, acmc-20dn5a, 5-tributylstannylpyrimidine, 5-tributylstannanyl-pyrimidine, tri-n-butyl 5-pyrimidinyl tin, 5-tri-butylstannyl pyrimidine, tributyl 5-pyrimidinyl stannane
PubChem CID 2763249
IUPAC Name tributyl(pyrimidin-5-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CN=CN=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15438097
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H51458.MD
250 mg
79.60 EUR
250mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15428097
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H51458.03
1 g
246.00 EUR
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

144173-85-3
369.14
QRDQHTJNKPXXRQ-UHFFFAOYSA-N
2763249
CCCC[Sn](CCCC)(CCCC)C1=CN=CN=C1
C16H30N2Sn
MFCD01319056
5-tributylstannyl pyrimidine, pyrimidine, 5-tributylstannyl, tributyl pyrimidin-5-yl stannane, 5-tri-n-butylstannyl pyrimidine, acmc-20dn5a, 5-tributylstannylpyrimidine, 5-tributylstannanyl-pyrimidine, tri-n-butyl 5-pyrimidinyl tin, 5-tri-butylstannyl pyrimidine, tributyl 5-pyrimidinyl stannane
tributyl(pyrimidin-5-yl)stannane

Specifications

144173-85-3
MFCD01319056
5-tributylstannyl pyrimidine, pyrimidine, 5-tributylstannyl, tributyl pyrimidin-5-yl stannane, 5-tri-n-butylstannyl pyrimidine, acmc-20dn5a, 5-tributylstannylpyrimidine, 5-tributylstannanyl-pyrimidine, tri-n-butyl 5-pyrimidinyl tin, 5-tri-butylstannyl pyrimidine, tributyl 5-pyrimidinyl stannane
CCCC[Sn](CCCC)(CCCC)C1=CN=CN=C1
369.14
369.13
5-(Tri-n-butylstannyl)pyrimidine
C16H30N2Sn
UN2788
QRDQHTJNKPXXRQ-UHFFFAOYSA-N
tributyl(pyrimidin-5-yl)stannane
2763249
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H372

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only