Learn More
Hamilton™ 50 μL 1700 Series Gas Tight Syringe
The 1700 Series syringes are the Gas Tight version of the 700 series syringes.
Brand:  Hamilton™ CAL80939
 View more versions of this product
Product Code. 17844342
  • 318.00 EUR / Each
Explore more special offers
Description
Description
  • This series of syringes are ideal for dispensing volumes up to 50 μL.
  • A variety of different terminations and needle options are available in this series.
Specifications
Specifications
Autoclavable
No Needle Included
0.305 in.
7.75 mm
Gas Tight Syringe
115°C
1705 TLL
1
No Needle Included
Luer Lock
PTFE
0.041 in.
1.03 mm
No Needle Included
239°F
50°F
10°C
50 μL
No Needle Included