Learn More
Hamilton™ 50 μL, Model 1705 TLL SYR, Long Life Instrument Syringe
The 1700 series syringes are the Gastight version of the original 700 series syringes. This series of syringes is ideal for dispensing volumes from 1 μL up to 500 μL. A variety of different terminations and needle options are available in this series.
Brand: Hamilton™ 8300-15
This item is not returnable.
View return policy
Description
Specification Sheet
Volume | 50 μL |
---|---|
Syringe Type | Longlife Gastight |
Syringe Series | 1700 |
Termination | PTFE Luer Lock (TLL) |
Needle | Instrument Syringe |
Plunger Style | X-Style |
Fluid Path | Borosilicate glass, PTFE, UHMWPE |
Autoclavable | No |
Gas Sterilizable | Yes |
Pressure | 1000 psig (68.9 bar) |
Maximum Temperature | 115°C / 239°F |
Minimum Temperature | 10°C / 50°F |
Barrel Inner Diameter | 0.041 inches (1.03 mm) |
Barrel Outer Diameter | 0.305 inches (7.75 mm) |
Sales Quantity | 1 syringe per pack |
Specifications
Longlife Gastight 1700 Series | |
50 μL |