Learn More
Tocris™ ML 418
Kir 7.1 inhibitor. Also inhibits Kir 6.2
Brand: Tocris™ 6889/50
This item is not returnable.
View return policy
Description
ML 418 is a Kir7.1 inhibitor (IC50 = 0.31 μM). Also inhibits Kir6.2 with similar potency. Displays >17-fold selectivity for Kir7.1 over Kir1.1, Kir2.1, Kir2.2, Kir2.3, Kir3.1/3.2 and Kir4.1. Active in vivo.
Specifications
Isopropyl (1-((5-chloro-8-hydroxyquinolin-7-yl)methyl)piperidin-4-yl)carbamate | |
50 mg | |
98% |
1928763-08-9 | |
C19H24CIN3O3 |
Product Content Correction
Product Title
Tocris™ ML 418 > 50 mg
Spot an opportunity for improvement?Share a Content Correction