Learn More
Spectrum™ Labs Spectra/Por™ 5 12-14 kD MWCO Standard RC Dry Dialysis Kits
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
Brand:  Spectrum™ 132754T
 View more versions of this product
Product Code. 15310782
  • 215.00 EUR / 5m
Explore more special offers
Includes: 5 m of Spectra/Por 5 dialysis tubing and 2 closures
Description
Description
  • For reinforced for weight applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry without glycerin (no soaking required)
Specifications
Specifications
12 to 14 kDa
48 mm
18 mL/cm vol./length
5 m Tubing/Kit