missing translation for 'onlineSavingsMsg'
Learn More
5alpha-Cholestan-3beta-ol, 98%, Thermo Scientific Chemicals Product Code.: 11382616

5alpha-Cholestan-3beta-ol, 98%, Thermo Scientific Chemicals

Product Code. 11382616
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11382616

Brand: Thermo Scientific Alfa Aesar L08624.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5alpha-Cholestan-3beta-ol is used as a standard in lipid analysis using HPLC. It acts as a derivitized steroid compound. It is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5alpha-Cholestan-3beta-ol is used as a standard in lipid analysis using HPLC. It acts as a derivitized steroid compound. It is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-97-7
Molecular Formula C27H48O
Molecular Weight (g/mol) 388.68
MDL Number MFCD00066413
InChI Key QYIXCDOBOSTCEI-UHFFFAOYNA-N
Synonym coprostan-3alpha-ol
PubChem CID 44135456
IUPAC Name (10S,13R)-10,13-dimethyl-17-(6-methylheptan-2-yl)-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC(C)CCCC(C)C1CCC2C3CCC4CC(O)CCC4(C)C3CCC12C

Specifications

Melting Point 140°C to 144°C
Quantity 5 g
Beilstein 2418594
Merck Index 14,2200
Solubility Information Insoluble in water.
Optical Rotation +24° (c=1 in Chloroform)
Formula Weight 388.68
Percent Purity 98%
Chemical Name or Material 5alpha-Cholestan-3beta-ol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10246713 Certificate TypeCertificate of Analysis Date22/05/2023 Catalog NumberL08624.06,L08624.14
Lot Number10237955 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL08624.06,L08624.14
Lot Number10242892 Certificate TypeCertificate of Analysis Date16/08/2022 Catalog NumberL08624.06,L08624.14
Lot Number10240104 Certificate TypeCertificate of Analysis Date08/04/2022 Catalog NumberL08624.06,L08624.14
Lot Number10232425 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL08624.06,L08624.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5alpha-Cholestan-3beta-ol, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.