missing translation for 'onlineSavingsMsg'
Learn More

V-Solv™ ICP Solvent, VHG™

Product Code. 17940806
Click to view available options
Quantity:
1 gal
5 gal
200 L
Unit Size:
1 GAL
200 L
5 GAL
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 17940806

Brand: VHG Labs VHGVSOLV5GAL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

V-Solv™ ICP Solvent
TRUSTED_SUSTAINABILITY

Specifications

Methods ICP
Shipping Condition Room Temperature
Solvent or Matrix V-SOLV™ ICP Solvent
Quantity 5 gal
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.