missing translation for 'onlineSavingsMsg'
Learn More

6-(4-Boc-1-piperazinyl)pyridine-3-boronic acid pinacol ester, 99%, Thermo Scientific™

Product Code. 11459260
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496786-98-2
Molecular Formula C20H32BN3O4
Molecular Weight (g/mol) 389.30
MDL Number MFCD04039875
InChI Key JWUBVPJWWYYRLJ-UHFFFAOYSA-N
Synonym tert-butyl 4-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl piperazine-1-carboxylate, 4-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-yl-piperazine-1-carboxylic acid tert-butyl ester, 1-boc-4-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl piperazine, 6-4-boc-1-piperazinyl pyridine-3-boronic acid pinacol ester, 2-4-boc-piperazine pyridine-5-boronic acid pinacol ester, 2-4-n-boc piperazin-1-yl pyridine-5-boronic acid pinacol ester, 2-4-tert-butoxycarbonylpiperazin-1-yl pyridine-5-boronic acid, pinacol ester, tert-butyl 4-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl piperazine-1-carboxylate, 2-4-boc-piperazino pyridine-5-boronic acid pinacol ester
PubChem CID 16217947
IUPAC Name tert-butyl 4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(C=N1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 250 mg
Formula Weight 389.31
Percent Purity 99%
Chemical Name or Material 6-(4-Boc-1-piperazinyl)pyridine-3-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'