Learn More
6-(Benzyloxy)-2-naphthylboronic acid, 97%, Thermo Scientific™
35.40 EUR - 314.00 EUR
Chemical Identifiers
CAS | 152915-83-8 |
---|---|
Molecular Formula | C17H15BO3 |
Molecular Weight (g/mol) | 278.114 |
MDL Number | MFCD04115402 |
InChI Key | ZVHAZMUBLCEMGG-UHFFFAOYSA-N |
Synonym | 6-benzyloxy-2-naphthylboronic acid, 6-benzyloxynaphthalene-2-boronic acid, 6-benzyloxy naphthalen-2-ylboronic acid, boronic acid,b-6-phenylmethoxy-2-naphthalenyl, 6-benzyloxy-2-naphthaleneboronic acid, 6-benzyloxy naphthalen-2-yl boronic acid, 6-phenylmethoxynaphthalen-2-yl boronic acid, acmc-209d8c, 6-benzyloxy-2-naphthylboronicacid |
PubChem CID | 2794628 |
IUPAC Name | (6-phenylmethoxynaphthalen-2-yl)boronic acid |
SMILES | B(C1=CC2=C(C=C1)C=C(C=C2)OCC3=CC=CC=C3)(O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10394672
|
Thermo Scientific Maybridge
MO00847CB |
250 mg | Amber Glass Bottle |
35.40 EUR
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10304682
|
Thermo Scientific Maybridge
MO00847DA |
1 g | Amber Glass Bottle |
90.20 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10206004
|
Thermo Scientific Maybridge
MO00847DE |
5 g | Amber Glass Bottle |
314.00 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
152915-83-8 | |
278.114 | |
ZVHAZMUBLCEMGG-UHFFFAOYSA-N | |
2794628 | |
B(C1=CC2=C(C=C1)C=C(C=C2)OCC3=CC=CC=C3)(O)O |
C17H15BO3 | |
MFCD04115402 | |
6-benzyloxy-2-naphthylboronic acid, 6-benzyloxynaphthalene-2-boronic acid, 6-benzyloxy naphthalen-2-ylboronic acid, boronic acid,b-6-phenylmethoxy-2-naphthalenyl, 6-benzyloxy-2-naphthaleneboronic acid, 6-benzyloxy naphthalen-2-yl boronic acid, 6-phenylmethoxynaphthalen-2-yl boronic acid, acmc-209d8c, 6-benzyloxy-2-naphthylboronicacid | |
(6-phenylmethoxynaphthalen-2-yl)boronic acid |
Specifications
152915-83-8 | |
97% | |
MFCD04115402 | |
ZVHAZMUBLCEMGG-UHFFFAOYSA-N | |
(6-phenylmethoxynaphthalen-2-yl)boronic acid | |
2794628 | |
97% |
97.0 | |
C17H15BO3 | |
6-benzyloxy-2-naphthylboronic acid, 6-benzyloxynaphthalene-2-boronic acid, 6-benzyloxy naphthalen-2-ylboronic acid, boronic acid,b-6-phenylmethoxy-2-naphthalenyl, 6-benzyloxy-2-naphthaleneboronic acid, 6-benzyloxy naphthalen-2-yl boronic acid, 6-phenylmethoxynaphthalen-2-yl boronic acid, acmc-209d8c, 6-benzyloxy-2-naphthylboronicacid | |
B(C1=CC2=C(C=C1)C=C(C=C2)OCC3=CC=CC=C3)(O)O | |
278.114 | |
278.11 | |
6-(Benzyloxy)-2-naphthylboronic acid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.