missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-iodo-3-hydroxypyridine, 98%, Thermo Scientific Chemicals

Product Code. 15405927
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2,2'-Dipyridylamine has been obtained by by heating a mixture of 2-aminopyridine and 2-bromo- (chloro-, iodo- ) -pyridine with anhydrous zinc chloride or with barium oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Dipyridylamine has been obtained by by heating a mixture of 2-aminopyridine and 2-bromo- (chloro-, iodo- ) -pyridine with anhydrous zinc chloride or with barium oxide.

Solubility
Slightly Soluble (1.3 g/L) (25°C).

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188057-26-3
Molecular Formula C5H3ClINO
Molecular Weight (g/mol) 255.439
MDL Number MFCD07636749
InChI Key FWIMPBYPMQSSCD-UHFFFAOYSA-N
Synonym 6-chloro-2-iodo-3-hydroxypyridine, 6-chloro-2-iodo-3-pyridinol, 6-chloro-2-iodo-pyridin-3-ol, 3-pyridinol, 6-chloro-2-iodo, 2-iodo-6-chloropyridine-3-ol, 3-pyridinol,6-chloro-2-iodo
PubChem CID 23433158
IUPAC Name 6-chloro-2-iodopyridin-3-ol
SMILES C1=CC(=NC(=C1O)I)Cl

Specifications

Melting Point 191°C to 193°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Slightly Soluble (1.3g/L) (25°C).
Formula Weight 255.44
Percent Purity 98%
Chemical Name or Material 6-Chloro-2-iodo-3-hydroxypyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'