missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 6-FAM (6-Carboxyfluorescein), single isomer

Product Code. 11554227
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 11554227

Brand: Invitrogen™ C1360

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

6-FAM (6-Carboxyfluorescein), single isomer

The single isomer, 6-FAM, contains a carboxylic acid that can be used to react with primary amines via carbodiimide activation of the carboxylic acid. Fluorescein is the most common fluorescent derivatization reagent for labeling biomolecules. In addition to its relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility, fluorescein has an excitation maximum that closely matches the 488 nm spectral line of the argon-ion laser.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mg
Product Type 6-FAM
Content And Storage Store at room temperature and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Carboxylic Acid
Label or Dye FITC (Fluorescein)
Label Type Classic Dyes
Excitation 488

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.