Learn More
Thermo Scientific™ 8 x 8 PCR Strip Fixed Angle Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
Shop All Thermo Scientific ProductsDescription
Support micro-volume protocols, such as PCR reaction set-up, with the Thermo Scientific™ 8 x 8 PCR Strip Fixed Angle Rotor: Capable of spinning up to 15,000rpm / 17,860 x g for your PCR processing needs.
Supports micro-volume protocols, such as PCR reaction set-up, complete with our innovative Thermo Scientific technologies.
Secure, push-button Auto-Lock rotor exchange in less than 3 seconds delivers:
- Trouble-free rotor installation and removal
- No tools are required
- No complicated rotor changing procedures
- Clear chamber access for cleaning convenience
- Quick switch between applications, from conical tubes to blood tubes, from microtubes to microplates, and the flexibility to evolve with the changing needs of your laboratory.
Certified† biocontainment rotor lid for glove-friendly, one-handed operation.
Ordering Information:
Replacement biocontainment lid (75005730) and replacement O-rings (75005726) are sold separately. Contact your sales representative for more information.
Specifications
Specifications
| Description | 8 x 8 PCR Strip Fixed Angle Rotor |
| Type | Fixed Angle Rotor |
| Capacity | 8 x 8 PCR Strips |
| Max. RCF | 17,860 x g |
| Max. Speed | 15,000 rpm |
| Radius (Metric) | 71 mm |
| K Factor | 553 |
| For Use With (Equipment) | Thermo Scientific Sorvall ST 8, Megafuge 8 and SL 8 Centrifuges |
| Angle | 45° |
| Biocontainment | Yes |
† Biocontainment certification by the Public Health England, Porton Down, UK.