missing translation for 'onlineSavingsMsg'
Learn More

9-Ethynyl-9-fluorenol, Thermo Scientific™

Product Code. 10137642 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10137642

Brand: Thermo Scientific Acros 381540050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13461-74-0
MDL Number MFCD00041555
InChI Key MMZVVJGCZZAWBN-UHFFFAOYSA-N
Synonym 9-ethynyl-9-fluorenol, 9-ethynyl-9h-fluoren-9-ol, acmc-209bwd, 9-ethynyl-9-hydroxyfluorene, 9-ethinyl-9-hydroxy-fluorene, 9h-fluoren-9-ol,9-ethynyl, 9-ethynyl-9h-fluoren-9-ol #, mmzvvjgczzawbn-uhfffaoysa, 9-hydroxy-9-fluorenyl acetylene, 9-ethynyl-9-fluorenol,
PubChem CID 518771
IUPAC Name 9-ethynylfluoren-9-ol
SMILES C#CC1(C2=CC=CC=C2C3=CC=CC=C31)O

Specifications

Melting Point 107°C to 110°C
Color White
Infrared Spectrum Authentic
Packaging Glass Bottle
Molecular Formula C15H10O
Quantity 5 g
Molecular Weight (g/mol) 206.244
Formula Weight 206.24
Percent Purity ≥98.5%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 9-Ethynyl-9-fluorenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.