Learn More
uvex 9169 Super F Over-the-Glasses Safety Glasses
These extremely comfortable safety spectacles provide excellent eye area protection, and fit perfectly over conventional prescription lenses. Specs are available in a variety of lens coating and lens tints.
19.85 EUR - 22.50 EUR
Specifications
Model | 9169 Super F OTG |
---|---|
Material | Polycarbonate |
Disposable | Reusable |
Lens Material | Polycarbonate |
Frame Material | Plastic |
Product Code | Brand | Lens Coating | Lens Tint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Lens Coating | Lens Tint | Price | Quantity & Availability | ||||
17217261
|
uvex
9169261 |
Anti-fog, Scratch-resistant | Clear |
19.85 EUR
Each |
Estimated Shipment: 13-11-2024
Log in to see stock. |
||||
17207261
|
uvex
9169164 |
Anti-fog, Scratch-resistant | Light Brown |
22.50 EUR
Each |
Estimated Shipment: 13-11-2024
Log in to see stock. |
||||
17237261
|
uvex
9169580 |
Scratch-resistant | Amber | ||||||
Description
- Overspecs for users who also wear prescription lenses with an arm-hinge concept
- Side arms graduate to soft, flexible material for enhanced comfort and grip
- Unrestricted peripheral vision
Supravision Excellence Coating
- Inside: Anti-fog
- Outside: Scratch-resistant and chemical-resistant
- Easy to clean, and less susceptible to soiling
Supravision Sapphire Coating
- Inside: Scratch-resistant and chemical-resistant
- Outside: Scratch-resistant and chemical-resistant
- Non-stick properties allow for easy removal of greasy/watery dirt
Infradur Plus Coating
- Inside: Anti-fog
- Outside: Scratch-resistant
- Minimizes burns due to welding sparks
Specifications
9169 Super F OTG | |
Reusable | |
Plastic |
Polycarbonate | |
Polycarbonate | |
Standard |