Learn More
TPP™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Product Code. 11331774
Click to view available options
Packaging:
1 Pk., 162 Cases
6/Pk., 108/Cs.
Color: Clear; Sterility; Sterile; Lid With Lid
Color: Clear; Sterility; Sterile; Lid With Lid
  • Air-venting system in lid guarantees controlled gas exchange with low evaporation; vents in plate base prevent condensation and provide consistent air-flow between stacked plates
  • Sloped edge allows placement of the lid in one position only;  yellow marking area on lid and plate ensure correct lid orientation
  • Ridged grip area ensures a secure grasp and prevents accidental lifting of lid
  • Easy to see black alpha-numerical labelling between the wells for easy orientation during microscopy
  • Growth area on spherical zone only; absolutely flat growth surface
  • Plate sizes (6, 12, 24, 48 and 96-well) stack securely together
  • Manufactured meeting the standards ANSI/SLAS 1-2004 through ANSI/SLAS 4-2004
  • White base colour is embossed firmly on the test plate eliminating loss of identification through detachment
  • 2 well geometries available: F-base and U-base
Specifications
Barcode Alphanumeric
Format Standard
Lid With Lid
Culture Area 0.322 cm2
Array 8 x 12
Packaging 6/Pk., 108/Cs.
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
Quantity 1/Bag
Dimensions (L x W x H) 128 x 86 x 17 mm