Learn More
Thermo Scientific™ 96-Well Flipper™ Microtube Racks
Shop All MBP Molecular BioProducts Products
Click to view available options
Color:
Fluorescent Green
Fluorescent Orange
Fluorescent Pink
Red
Unit Size:
Pack of 10
Description
Protect samples and allow racks to be stacked using the Thermo Scientific™ 96-Well Flipper™ Microtube Rack. The rack, which is molded with an alphanumeric grid for easy tube identification, holds 0.5mL tubes on one side and 1.5mL tubes on the other.
- Holds 0.5mL tubes on one side, 1.5mL tubes on the other
- Molded with an alphanumeric grid for easy tube identification
- Several colors available
Specifications
Specifications
| Product Type | 96-well Flipper Rack |
| Description | Flipper Rack |
| Color | Fluorescent Orange |
| Holds | Microtubes (2 mL), Microtubes (1.5 mL), Microtubes (2 mL) |
| Material | Polypropylene |
| Shape | Rectangular |
| Stackable | Yes |
| No. per Case | 10 Racks |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur