missing translation for 'onlineSavingsMsg'
Learn More

Green Elephant 96-Well Plate

Green Elephant 96-well plate comes with a 50% reduced carbon footprint. The microtiter plate is made from renewable biopolymer polylactic acid (PLA) and provides great functionality and quality. The intransparent design is ideal for fluorescence-based assays and screenings.

Brand:  Green Elephant GEB-WP96FBT

 View more versions of this product

Product Code. 30019058

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Flat-bottom, clearly labeled wells for easy identification
  • Made from plant-based biopolymer polylactic acid (PLA)
  • Improving sustainability by a 50% reduced carbon footprint
  • Great optical features for microscopic applications and photometric measurements
  • Compatibility with robotics, plate readers, and liquid handling systems
  • Temperature resistant between -80 °C and +50 °C
  • High mechanical and broad chemical resistance
  • Centrifugation stability up to 4000 x g
Specifications

Specifications

No Barcode
No Lid
Not Autoclavable
Single-use
Polymer
96
Non-sterile
Made from renewable biopolymer polylactic acid (PLA)
100/Cs.
SBS Standard
8 x 12
Transparent, Clear Bottom
Microtiter Plate
372 μL
50 to 200 μL
Flat Bottom (F-well)
100 Plates
127.76 x 85.18 x 14.35 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.