missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Green Elephant 96-Well Plate
Green Elephant 96-well plate comes with a 50% reduced carbon footprint. The microtiter plate is made from renewable biopolymer polylactic acid (PLA) and provides great functionality and quality. The intransparent design is ideal for fluorescence-based assays and screenings.
Brand: Green Elephant GEB-WP96FBT
This item is not returnable.
View return policy
Description
- Flat-bottom, clearly labeled wells for easy identification
- Made from plant-based biopolymer polylactic acid (PLA)
- Improving sustainability by a 50% reduced carbon footprint
- Great optical features for microscopic applications and photometric measurements
- Compatibility with robotics, plate readers, and liquid handling systems
- Temperature resistant between -80 °C and +50 °C
- High mechanical and broad chemical resistance
- Centrifugation stability up to 4000 x g
Specifications
No Barcode | |
No Lid | |
Not Autoclavable | |
Single-use | |
Polymer | |
96 | |
Non-sterile | |
Made from renewable biopolymer polylactic acid (PLA) | |
100/Cs. |
SBS Standard | |
8 x 12 | |
Transparent, Clear Bottom | |
Microtiter Plate | |
372 μL | |
50 to 200 μL | |
Flat Bottom (F-well) | |
100 Plates | |
127.76 x 85.18 x 14.35 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction