Learn More
Eppendorf™ twin.tec™ PCR Plates LoBind™ - PCR Plates
GREENER_CHOICE
Product Code. 15861525 Shop All Eppendorf Products
Click to view available options
Color:
Blue
Clear
Green
Orange
Red
Yellow
No. of Wells:
96
384
Volume (Metric) Well:
45 μL
150 μL
250 μL
Packaging:
10 Bags x 30 Plates
300 Plates
5 Bags x 5 Plates
Unit Size:
Pack of 25
Pack of 300
Product Code. 15861525
Brand: Eppendorf™ 0030129555
Only null left
Add to basket
Add to basket
Eppendorf twin.tec™ PCR plates LoBind, available in 96- and 384-well formats, maximize nucleic acid recovery and are certified as PCR clean for consistently reliable results.
  • LoBind material guarantees maximum sample recovery for improved assay results.
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • OptiTrack™ matrix for faster sample identification and fewer pipetting errors.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean).
TRUSTED_SUSTAINABILITY
Specifications
Barcode Includes Barcode
Binding Property Low DNA Binding
Certifications/Compliance PCR Clean
Certified Yes
Format Standard
Lid With Lid
Array 8 x 12
Autoclavable Autoclavable
Packaging 10 Bags x 30 Plates
Product Line Eppendorf
Color Green
Disposable Single-use
Product Type PCR Plate
Skirt Style Skirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 150 μL
No. of Wells 96
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
For Use With (Application) Real Time PCR
High-throughput Compatibility Yes
Max. RCF 6000 x g
No. per Pack 300 Pk.
Plate Compatibility Yes
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 300 Pk.
Rimmed Yes
Well Color Colorless