missing translation for 'onlineSavingsMsg'
Learn More

A-867744, MedChemExpress

MedChemExpress A-867744 is a highly potent and selective type II positive allosteric modulator (PAM) of the alpha7 nicotinic acetylcholine receptors (nAChR) with an EC50 of 1.0 μM.

166.05 EUR - 996.30 EUR

Specifications

Chemical Name or Material A-867744
CAS 1000279-69-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H19ClN2O3S
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16481237
Encompass_Preferred
View Documents
Medchem Express
HY-12149
50 mg
557.55 EUR
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16465756
Encompass_Preferred
View Documents
Medchem Express
HY-12149
100 mg
996.30 EUR
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16444998
Encompass_Preferred
View Documents
Medchem Express
HY-12149
10 mg
166.05 EUR
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A-867744 is a highly potent and selective type II positive allosteric modulator (PAM) of the alpha7 nicotinic acetylcholine receptors (nAChR) with an EC50 of 1.0 μM.
Specifications

Specifications

A-867744
Off-White
C20H19ClN2O3S
O=S(C1=CC=C(C=C1)N2C(C)=C(C=C2C3=CC=C(C=C3)Cl)C(CC)=O)(N)=O
402.89
Research
Neuroscience-Neuromodulation
1000279-69-5
Research
DMSO : ≥ 100 mg/mL (248.21 mM)
402.89
95.06%
Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.