Learn More
Thermo Scientific™ Abgene Blue Racks
GREENER_CHOICE
Product Code. 11954714
Click to view available options
Color:
Blue
Unit Size:
Pack of 10
Product Code. 11954714
Brand: Thermo Scientific™ AB1434
Only null left
Add to basket
Add to basket
The Thermo Scientific Abgene 2D Barcoded 2mL Screw Cap storage tubes combine a traditional storage format with the added benefits of a secure sample tracking and a high density, microplate footprint rack for easier sample handling.
Secure Tracking
  • 2D barcode- is visible- even in the harshest of laboratory conditions
  • Instantly decode each tube's 2D barcode with the Thermo Scientific line of 2D barcode readers; scan the tube into any application or database, or integrate via various data export options
  • Once sample data is stored, scan the tube using the database Find feature to instantly access sample information and identify the contents of each storage tube
Superior Design
  • Save precious freezer space with an innovative, stackable, 6 x 8 microplate-footprint rack of 48 tubes with a familiar 2mL cryopreservation format
  • Alphanumeric grid-referencing on rack and lid clearly display the storage tube location
  • Externally threaded tubes prevent contamination
  • Open rack bottom allows all barcodes to be read at once, saving time versus and allowing more efficient freezing and thawing
  • Unlike other plastic or cardboard storage boxes, the stackable, secure rack also ensures your freezer stays organised for faster sample retrieval
  • V-shaped internal well design reduces dead volume and improves sample recovery
Stringent Quality Control
  • Each tube is scanned to ensure readability
  • Each code is checked against a full database of all previously assigned 2Dcodes to ensure zero duplicates across the entire line of Abgene 2D barcoded storage tubes
  • Each lot of storage tubes is leak tested to ensure the integrity and security of samples
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
For Use With (Equipment) 2mL 2D barcoded tubes