Learn More
Lonza™ ABM™ Astrocyte Basal Medium
Unsupplemented basal medium designed to support the growth of primary human astrocytes.
Brand: Lonza™ CC-3187
This item is not returnable.
View return policy
Description
- Basal medium.
- Contains no growth factors cytokines, or supplements.
- This product is available in a BulletKit™ consisting of basal medium and SingleQuots™ growth factor supplements, which are also available separately.
- Developed to support the growth of primary human astrocytes.
Specifications
Red | |
Astrocytes | |
7.6 to 8.0 |
Store in dark at 2 to 8°C | |
Basal Medium | |
500 mL |