Learn More
BINDER™ Left Access Port (50 mm)
Material: Silicone; For Use With: BINDER units KB 240, KB 400, KB 720
Brand: BINDER™ 8012-0653
This item is not returnable.
View return policy
Description
- For use on left
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Access port with silicone plug - 50 mm, left | |
Silicone | |
Silicone plug |
BINDER Chambers KB 240, KB 400, KB 720 | |
Plug | |
50 mm |