missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Accessories for G20 Titrator Models
For use with G20 Titrator models.
Brand: Mettler Toledo™ 51109828
This item is not returnable.
View return policy
Description
- Made of Hastelloy C22
- For temperature range 0° to 130°C
- With phototrode DP5
Specifications
Temperature Sensor | |
Hastelloy C22 | |
0°C to 130°C |
Phototrode DP5 | |
0°F to 266°F |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction