Learn More
Sartorius Accessories and Replacement Parts for Balances: Switches
GREENER_CHOICE
Product Code. 10110751 Shop All Sartorius Products
Click to view available options
For Use With (Equipment):
CPA Balances, Extended Balances, Master Pro LA Balances
Draft Shield of SE and ME Ultramicro and Microbalance
Type:
Foot Switch
Hand Switch
Unit Size:
Each
Product Code. 10110751
Brand: Sartorius YFS01
Balance Switches for remote control.
  • Hand and foot switch accessories for Sartorius balances
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) CPA Balances, Extended Balances, Master Pro LA Balances
Includes T-connector
Type Foot Switch