Learn More
Miele Professional™ Accessory large washer-disinfector
Plinth SBWR/2, on wheels including drip tray without openings for connecting services to G 7825
Brand:  Miele Professional™ 5653130
Product Code. 15440525
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
  • For models with drain pump
  • Castors for pulling the machine forward when in service
  • Installation of the supply hoses from above
  • Easy access when in service
Specifications
Specifications
Stainless Steel
Plinth/base
Stainless Steel
Large-capacity washer-disinfectors, laboratory