Learn More
Miele Professional™ Accessory large washer-disinfector
Magnetic strip ML/2 for automatic recognition of mobile unit/spray arm monitoring
Brand: Miele Professional™ 5723270
This item is not returnable.
View return policy
Description
- ML/2 is required for spray arm monitoring
- 5 magnets, individually configurable
- 15 possible combinations
Specifications
Stainless Steel | |
Magnetic Strip for Automatic Mobile Unit Recognition | |
Stainless Steel, Black | |
PG 8535, PG 8536 |