missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACE/CD143 Mouse anti-Human, Clone: 9B9, Novus Biologicals™
Mouse Monoclonal Antibody
253.00 EUR
Specifications
Antigen | ACE/CD143 |
---|---|
Clone | 9B9 |
Concentration | 0.2 mg/ml |
Dilution | ELISA |
Applications | ELISA |
Description
ACE/CD143 Monoclonal specifically detects ACE/CD143 in Human samples. It is validated for ELISA.Specifications
ACE/CD143 | |
0.2 mg/ml | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA | |
Full-length recombinant human ACE/CD143 protein | |
Primary | |
Store at 4°C. |
9B9 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
1636 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title