Learn More
Acebutolol hydrochloride, MedChemExpress
Click to view available options
Quantity:
100 mg
5 g
10 g
Unit Size:
100mg
10g
5g
Description
Acebutolol hydrochloride is an orally active β1 adrenergic receptor (β1AR) antagonist. Acebutolol hydrochloride is used in the treatment of hypertension, angina pectoris and cardiac arrhythmias.
Specifications
Specifications
| Chemical Name or Material | Acebutolol hydrochloride |
| CAS | 34381-68-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H29ClN2O4 |
| Quantity | 5 g |
| Solubility Information | DMSO : 100 mg/mL (268.18 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (134.09 mM) |
| SMILES | CCCC(NC1=CC=C(OCC(O)CNC(C)C)C(C(C)=O)=C1)=O.Cl |
| Molecular Weight (g/mol) | 372.89 |
| Formula Weight | 372.89 |
| Show More |