missing translation for 'onlineSavingsMsg'
Learn More

Acebutolol hydrochloride, MedChemExpress

MedChemExpress Acebutolol hydrochloride is an orally active β1 adrenergic receptor (β1AR) antagonist. Acebutolol hydrochloride is used in the treatment of hypertension, angina pectoris and cardiac arrhythmias.

Brand:  Medchem Express HY-17497A

Encompass_Preferred

 View more versions of this product

Product Code. 16437368

  • POA / 10g

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Acebutolol hydrochloride is an orally active β1 adrenergic receptor (β1AR) antagonist. Acebutolol hydrochloride is used in the treatment of hypertension, angina pectoris and cardiac arrhythmias.
Specifications

Specifications

Acebutolol hydrochloride
White
C18H29ClN2O4
DMSO : 100 mg/mL (268.18 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (134.09 mM)
372.89
98.83%
Powder
34381-68-5
Research
10 g
CCCC(NC1=CC=C(OCC(O)CNC(C)C)C(C(C)=O)=C1)=O.Cl
372.89
Research
COVID-19-immunoregulation
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research purposes only