Learn More
ACETATE BUFFER SOLUTION PH 4.65, SODIUM ACETATE / ACETIC ACID, Honeywell Fluka™
Product Code. p-7122051 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
Pack of 6
Product Code. 15623220
Brand: Honeywell-Fluka 311031L
Request Bulk or Custom Format
SODIUM ACETATE / ACETIC ACID
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Specifications
CAS 127-09-3
Quantity 1 L
Molecular Formula C2H3NaO2
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
SMILES [Na+].CC([O-])=O
IUPAC Name sodium acetate
Molecular Weight (g/mol) 82.03