missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, glacial, ≥99%, MP Biomedicals™
Click to view available options
Quantity:
500 g
1 kg
Specifications
Specifications
Assay Percent Range | ≥99% |
Formula Weight | 60.05 |
Density | 1.049g/cm3 at 20°C |
Quantity | 500 g |
Odor | Very Pungent |
Physical Form | Liquid |
Chemical Name or Material | Acetic acid, glacial |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction