Learn More
Acetic acid, glacial, ≥99%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
1 kg
Specifications
Specifications
Assay Percent Range | ≥99% |
Formula Weight | 60.05 |
Density | 1.049g/cm3 at 20°C |
Quantity | 1 kg |
Odor | Very Pungent |
Physical Form | Liquid |
Chemical Name or Material | Acetic acid, glacial |