missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid glacial, HPLC grade, Scharlab
Click to view available options
Quantity:
1 L
Specifications
Specifications
CAS | 64-19-7 |
Molecular Formula | C2H4O2 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
SMILES | CC(O)=O |
IUPAC Name | acetic acid |
Molecular Weight (g/mol) | 60.05 |
Grade | HPLC |
Quantity | 1 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction