missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetoacetyl coenzyme A sodium salt, >90%, MP Biomedicals™
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Specifications
Specifications
Color | Yellow |
Assay Percent Range | >90% |
Quantity | 1 mg |
Formula Weight | 901.6 (free acid) |
Physical Form | Solid |
Chemical Name or Material | Acetoacetyl coenzyme A |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'