missing translation for 'onlineSavingsMsg'
Learn More
Acetone-1,3-dicarboxylic acid, 97%, Thermo Scientific Chemicals Product Code.: 11444774

Acetone-1,3-dicarboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 11444774
1000 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11444774

Brand: Thermo Scientific Alfa Aesar A13742.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Acetone-1,3-dicarboxylic acid is used as a building block in organic chemistry. It is also used in the synthesis of hetrocyclic rings and in the Weiss-Cook reaction, which involves the preparation of cis-bicyclo[3.3.0]octane-3,7-dione by reacting with diacyl (1,2 ketone). Its presence in human urine is used as a diagnostic test for the overgrowth of harmful gut flora such as Candida albicans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetone-1,3-dicarboxylic acid is used as a building block in organic chemistry. It is also used in the synthesis of hetrocyclic rings and in the Weiss-Cook reaction, which involves the preparation of cis-bicyclo[3.3.0]octane-3,7-dione by reacting with diacyl (1,2 ketone). Its presence in human urine is used as a diagnostic test for the overgrowth of harmful gut flora such as Candida albicans.

Solubility
Soluble in water and alcohol. Slightly soluble in ethyl acetate and ether. Insoluble in chloroform.

Notes
Hygroscopic. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-05-2
Molecular Formula C5H6O5
Molecular Weight (g/mol) 146.098
MDL Number MFCD00002711
InChI Key OXTNCQMOKLOUAM-UHFFFAOYSA-N
Synonym 1,3-acetonedicarboxylic acid, 3-oxoglutaric acid, acetonedicarboxylic acid, pentanedioic acid, 3-oxo, acetone-1,3-dicarboxylic acid, 3-ketoglutaric acid, unii-ih7p7wo21p, 1,3-acetonedicarboxylic acid; 3-ketoglutaric acid, ih7p7wo21p, acetondicarbonsaureShow MoreShow Less
PubChem CID 68328
IUPAC Name 3-oxopentanedioic acid
SMILES C(C(=O)CC(=O)O)C(=O)O

Specifications

Melting Point ∼132°C (decomposition)
Quantity 1000 g
Beilstein 1447081
Sensitivity Hygroscopic
Merck Index 14,68
Solubility Information Soluble in water and alcohol. Slightly soluble in ethyl acetate and ether. Insoluble in chloroform.
Formula Weight 146.1
Percent Purity 97%
Chemical Name or Material Acetone-1,3-dicarboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number5001Q12N Certificate TypeCertificate of Analysis Date11/04/2024 Catalog NumberA13742.18,A13742.0B,A13742.30
Lot Number10195836 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13742.18,A13742.0B,A13742.30
Lot Number10179690 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13742.18,A13742.0B,A13742.30
Lot NumberB17Q28 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13742.18,A13742.0B,A13742.30
Lot Number10108782 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13742.18,A13742.0B,A13742.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetone-1,3-dicarboxylic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.