missing translation for 'onlineSavingsMsg'
Learn More

Acetone cyanohydrin, 99%, stabilized, Thermo Scientific Chemicals

Product Code. 11357391
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-86-5
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.11
InChI Key MWFMGBPGAXYFAR-UHFFFAOYSA-N
Synonym acetone cyanohydrin, acetone cyanhydrin, 2-methyllactonitrile, 2-hydroxyisobutyronitrile, alpha-hydroxyisobutyronitrile, acetoncyanhydrin, 2-cyano-2-propanol, acetoncianidrina, acetonkyanhydrin, acetoncianhidrinei
PubChem CID 6406
ChEBI CHEBI:15348
IUPAC Name 2-hydroxy-2-methylpropanenitrile
SMILES CC(C)(C#N)O

Specifications

Melting Point -19.0°C
Color Colorless to Yellow
Density 0.9320g/mL
Boiling Point 82.0°C (23.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C(OH)CN
Refractive Index 1.3990 to 1.4010
Quantity 5 g
Beilstein 03,316
Fieser 01,5; 08,409; 11,1
Merck Index 15,66
Specific Gravity 0.932
Solubility Information Solubility in water: soluble.
Stabilizer Approx. 0.3% H3PO4
Viscosity 4.8 mPa.s (20°C)
Formula Weight 85.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Acetone cyanohydrin
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'