missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Dried, Reagent Grade, Scharlab

Product Code. 11309783
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Contains a very low water content of ≤0.01% H2. Acetone, Dried, Reagent Grade, Scharlab is a dry solvent for use in both organic and inorganic synthesis.

Reagent grade solvents are the most frequently used grade for laboratory analytical work. They correspond to a grade of high quality chemicals for laboratory and specialized industrial use.

Specifications

Melting Point -95°C
CAS 67-64-1
Color Colorless
Acidity Max. 0.0002meq/g
Alkalinity Max. 0.0002meq/g
Appearance Passes test
Density 0.79g/cm3 at 20°C
Boiling Point 56°C
Quantity 1 L
Flash Point −20°
Identification Passes test
Ignition Point 540°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3588
Residue after Evaporation Max. 0.0005%
Molecular Formula C3H6O
Linear Formula (CH3)2CO
MDL Number MFCD00008765
Vapor Pressure 233 hPa at 20°C
UN Number 1090
Water Max. 0.01%, Dried
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
PubChem CID 180
ChEBI CHEBI:15347
Formula Weight 58.08g/mol
Percent Purity Min. 99.8%
Grade Applications: solvents, analytical chemistry, synthesis of organic products, photography
Chemical Name or Material Acetone
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.